Search Thermo Fisher Scientific
Danger
H314 - Causes severe skin burns and eye damage
H331 - Toxic if inhaled
H272 - May intensify fire; oxidizer
H290 - May be corrosive to metals
EUH071 - Corrosive to the respiratory tract
P220 - Keep away from clothing and other combustible materials
P280 - Wear protective gloves/protective clothing/eye protection/face protection
P301 + P330 + P331 - IF SWALLOWED: Rinse mouth. Do NOT induce vomiting
P303 + P361 + P353 - IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water or shower
P304 + P340 - IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing
P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing
P310 - Immediately call a POISON CENTER or doctor/physician