Nitric acid, ACS reagent, 68-70% solution in water, Thermo Scientific Chemicals
Nitric acid, ACS reagent, 68-70% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nitric acid, ACS reagent, 68-70% solution in water, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
25 mL
2.5 L
Packaging:
Plastic bottle
Glass Bottle
Catalog number 424005001
Price (USD)/ Each
-
Quantity:
500 mL
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS7697-37-2
IUPAC Namenitric acid
Molecular FormulaHNO3
InChI KeyGRYLNZFGIOXLOG-UHFFFAOYSA-N
SMILESO[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid free from suspended matter or sediment
Color scale=<10 APHA
AppearanceMay darken during storage as a result of
Appearancephotochemical reactions
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only