Thermo Scientific Chemicals

Potassium oxalate monohydrate, 99%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 424020025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Potassium oxalate monohydrate, 99%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 424020025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 424020025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6487-48-5
IUPAC Name
dipotassium hydrate oxalate
Molecular Formula
C2H2K2O5
InChI Key
QCPTVXCMROGZOL-UHFFFAOYSA-L
SMILES
O.[K+].[K+].[O-]C(=O)C([O-])=O
Appearance (Color)
White
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Titration with KMnO4
98.5 to 101.0 %
Chloride (Cl)
=<20 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text