Thermo Scientific Chemicals

Perchloric Acid, ca. 70% Solution in Water, For analysis, Thermo Scientific Chemicals

Catalog number: 424030010
1 kg, Glass bottle, Each
Thermo Scientific Chemicals

Perchloric Acid, ca. 70% Solution in Water, For analysis, Thermo Scientific Chemicals

Catalog number: 424030010
1 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 424030010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7601-90-3
IUPAC Name
perchloric acid
Molecular Formula
ClHO4
InChI Key
VLTRZXGMWDSKGL-UHFFFAOYSA-N
SMILES
O[Cl](=O)(=O)=O
Chloride (Cl)
=<10 ppm
Sulfate (SO4)
=<10 ppm
Appearance (Form)
Clear liquid
Color scale
=<10 APHA
Titration with NaOH
69.0 to 72.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text