Thermo Scientific Chemicals

Phosphoric acid, ACS reagent, 85+% solution in water, Thermo Scientific Chemicals

Catalog number: 424045000
500 mL, EcoSafPak™ Glass Bottle, Each
Thermo Scientific Chemicals

Phosphoric acid, ACS reagent, 85+% solution in water, Thermo Scientific Chemicals

Catalog number: 424045000
500 mL, EcoSafPak™ Glass Bottle, Each
Quantity
Packaging
Catalog number: 424045000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7664-38-2
IUPAC Name
phosphoric acid
Molecular Formula
H3O4P
InChI Key
NBIIXXVUZAFLBC-UHFFFAOYSA-N
SMILES
OP(O)(O)=O
Insoluble matter
=<0.001 %
Heavy metals (as Pb)
=<10 ppm
Chloride (Cl)
=<3 ppm
Manganese (Mn)
=<0.5 ppm
Sulfate (SO4)
=<30 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text