Potassium ferricyanide, 99+%, ACS reagent
Potassium ferricyanide, 99+%, ACS reagent
Potassium ferricyanide, 99+%, ACS reagent
Potassium ferricyanide, 99+%, ACS reagent
Thermo Scientific Chemicals

Potassium ferricyanide, 99+%, ACS reagent

CAS: 13746-66-2 | C6FeK3N6 | 329.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
500 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number 424125000
Price (USD)
163.65
Special offer
Online exclusive
Ends: 26-Sep-2025
202.00
Save 38.35 (19%)
Each
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
163.65
Special offer
Online exclusive
Ends: 26-Sep-2025
202.00
Save 38.35 (19%)
Each
Add to cart
Chemical Identifiers
CAS13746-66-2
IUPAC Nametripotassium hexacyanoirontris(ylium)
Molecular FormulaC6FeK3N6
InChI KeyQMTKJUMXUDIUAQ-UHFFFAOYSA-N
SMILES[K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals and/or chunks
Titration Iodometric>=99.0 %
Appearance (Color)Orange to red
Insoluble matter=<0.005 %
Ferro compounds=<0.05 % as ferrocyanide ion ¢Fe(CN)6!4-
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Share catalog number, name or link