Potassium ferrocyanide trihydrate, 98.5%, ACS reagent
Potassium ferrocyanide trihydrate, 98.5%, ACS reagent
Potassium ferrocyanide trihydrate, 98.5%, ACS reagent
Potassium ferrocyanide trihydrate, 98.5%, ACS reagent
Thermo Scientific Chemicals

Potassium ferrocyanide trihydrate, 98.5%, ACS reagent

CAS: 14459-95-1 | C6H6FeK4N6O3 | 422.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
500 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number 424130025
Price (USD)
279.65
Online Exclusive
310.00
Save 30.35 (10%)
Each
-
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
279.65
Online Exclusive
310.00
Save 30.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS14459-95-1
IUPAC Nametetrapotassium hexacyanoironbis(ylium) trihydrate
Molecular FormulaC6H6FeK4N6O3
InChI KeyNLYKFUFIUWPPIZ-UHFFFAOYSA-N
SMILESO.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Insoluble matter=<0.005 %
Sulfate (SO4)Passes test (limit about 0.01%)
Chloride (Cl)=<100 ppm
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link