Thermo Scientific Chemicals

Potassium hydroxide, ca. 85%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 424140250
25 g, Each
Thermo Scientific Chemicals

Potassium hydroxide, ca. 85%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 424140250
25 g, Each
Quantity
Catalog number: 424140250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1310-58-3
IUPAC Name
potassium hydroxide
Molecular Formula
HKO
InChI Key
KWYUFKZDYYNOTN-UHFFFAOYSA-M
SMILES
[OH-].[K+]
Titration with HCl
>=85 % (KOH)
Potassium carbonate
=<2.0 % (K2CO3)
Sulfate (SO4)
=<0.003 %
Heavy metals (ICP-OES)
=<0.001 %
Iron (Fe)
=<0.001 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text