Potassium persulfate, 99+%, ACS reagent, Thermo Scientific Chemicals
Potassium persulfate, 99+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium persulfate, 99+%, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
500 g
Packaging:
Plastic bottle
Catalog number 424180025
Price (USD)/ Each
325.65
Online exclusive
383.00 
Save 57.35 (15%)
-
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
325.65
Online exclusive
383.00 
Save 57.35 (15%)
Add to cart
Potassium persulfate, 99+%, ACS reagent, Thermo Scientific Chemicals
Catalog number424180025
Price (USD)/ Each
325.65
Online exclusive
383.00 
Save 57.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namedipotassium [(sulfonatoperoxy)sulfonyl]oxidanide
Molecular FormulaK2O8S2
InChI KeyUSHAGKDGDHPEEY-UHFFFAOYSA-L
SMILES[K+].[K+].[O-]S(=O)(=O)OOS([O-])(=O)=O
Molecular Weight (g/mol)270.31
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with KMnO4>=99.0 %
Heavy metals (as Pb)=<0.001 %
Insoluble matter=<0.005 %
Chlorine compounds (as Cl)=<0.001 %
Iron (Fe)=<5 ppm
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Oxidation reagent
RUO – Research Use Only