Potassium phosphate, monobasic, 99+%, ACS reagent
Potassium phosphate, monobasic, 99+%, ACS reagent
Potassium phosphate, monobasic, 99+%, ACS reagent
Potassium phosphate, monobasic, 99+%, ACS reagent
Thermo Scientific Chemicals

Potassium phosphate, monobasic, 99+%, ACS reagent

CAS: 7778-77-0 | H2KO4P | 136.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 424200025
Price (USD)
257.65
Online Exclusive
286.00
Save 28.35 (10%)
Each
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
257.65
Online Exclusive
286.00
Save 28.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS7778-77-0
IUPAC Namepotassium dihydrogen phosphate
Molecular FormulaH2KO4P
InChI KeyGNSKLFRGEWLPPA-UHFFFAOYSA-M
SMILES[K+].OP(O)([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
Appearance (Form)Crystals
Titration with NaOH>=99.0 %
pH4.1 to 4.5 (5% soln. at 25°C)
Insoluble matter=<0.01 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link