Potassium sulfate, 99+%, for biochemistry
Potassium sulfate, 99+%, for biochemistry
Thermo Scientific Chemicals

Potassium sulfate, 99+%, for biochemistry

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Packaging:
View all
Plastic bottle
Catalog number 424210010
Price (USD)/ Each
174.00
-
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
174.00
Add to cart
Potassium sulfate, 99+%, for biochemistry
Catalog number424210010
Price (USD)/ Each
174.00
-
Add to cart
Chemical Identifiers
CAS7778-80-5
IUPAC Namedipotassium sulfate
Molecular FormulaK2O4S
InChI KeyOTYBMLCTZGSZBG-UHFFFAOYSA-L
SMILES[K+].[K+].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystals
Titration after Ion exchange>=99.0 %
Heavy metals (as Pb)=<5 ppm
Insoluble matter=<0.1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only