Potassium sulfate, 99+%, ACS reagent, Thermo Scientific Chemicals
Potassium sulfate, 99+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium sulfate, 99+%, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Catalog number 424222500
Price (USD)/ Each
48.65
Online exclusive
54.20 
Save 5.55 (10%)
-
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
48.65
Online exclusive
54.20 
Save 5.55 (10%)
Add to cart
Potassium sulfate, 99+%, ACS reagent, Thermo Scientific Chemicals
Catalog number424222500
Price (USD)/ Each
48.65
Online exclusive
54.20 
Save 5.55 (10%)
-
Add to cart
Chemical Identifiers
CAS7778-80-5
IUPAC Namedipotassium sulfate
Molecular FormulaK2O4S
InChI KeyOTYBMLCTZGSZBG-UHFFFAOYSA-L
SMILES[K+].[K+].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration after Ion exchange>=99.0 %
Heavy metals (as Pb)=<5 ppm
Insoluble matter=<0.01 %
Nitrogen compounds (as N)=<5 ppm
pH5.5 to 8.5 (5% soln. at 25°C)
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only