Potassium thiocyanate, 99%, ACS reagent, Thermo Scientific Chemicals
Potassium thiocyanate, 99%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium thiocyanate, 99%, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 424230050
Price (USD)/ Each
40.65
Online exclusive
44.80 
Save 4.15 (9%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
40.65
Online exclusive
44.80 
Save 4.15 (9%)
Add to cart
Potassium thiocyanate, 99%, ACS reagent, Thermo Scientific Chemicals
Catalog number424230050
Price (USD)/ Each
40.65
Online exclusive
44.80 
Save 4.15 (9%)
-
Add to cart
Chemical Identifiers
CAS333-20-0
IUPAC Namepotassium cyanosulfanide
Molecular FormulaCKNS
InChI KeyZNNZYHKDIALBAK-UHFFFAOYSA-M
SMILES[K+].[S-]C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only