Thermo Scientific Chemicals

Acetic acid, sodium salt trihydrate, 99+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 424260250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

Acetic acid, sodium salt trihydrate, 99+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 424260250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 424260250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
6131-90-4
IUPAC Name
sodium acetate trihydrate
Molecular Formula
C2H9NaO5
InChI Key
AYRVGWHSXIMRAB-UHFFFAOYSA-M
SMILES
O.O.O.[Na+].CC([O-])=O
Infrared spectrum
Conforms
Titration with HClO4
99.0 to 101.0 %
Heavy metals (as Pb)
=<5 ppm
Potassium (K)
=<0.005 %
Sulfate (SO4)
=<0.002 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text