Thermo Scientific Chemicals

Sodium bicarbonate, 99.7+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 424270010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Sodium bicarbonate, 99.7+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 424270010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 424270010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
144-55-8
IUPAC Name
sodium hydrogen carbonate
Molecular Formula
CHNaO3
InChI Key
UIIMBOGNXHQVGW-UHFFFAOYSA-M
SMILES
[Na+].OC([O-])=O
Appearance (Color)
White
Appearance (Form)
Powder
Titration with HCl
99.7 to 100.3 % (on dried basis)
Heavy metals (ICP-OES)
=<5 ppm
Insoluble matter
=<0.015 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text