Thermo Scientific Chemicals

Sodium chloride, 99+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 424290251
25 kg, Fibre drum, Each
Thermo Scientific Chemicals

Sodium chloride, 99+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 424290251
25 kg, Fibre drum, Each
Quantity
Packaging
Catalog number: 424290251
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7647-14-5
IUPAC Name
sodium chloride
Molecular Formula
ClNa
InChI Key
FAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES
[Na+].[Cl-]
Titration Argentometric
>=99.0 %
Heavy metals (ICP-OES)
=<5 ppm
Insoluble matter
=<0.005 %
Iodide (I)
=<0.002 %
Chlorate and Nitrate (as NO3)
=<0.003 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text