Thermo Scientific Chemicals

Diethyldithiocarbamic acid, sodium salt trihydrate, ACS reagent, Thermo Scientific Chemicals

Catalog number: 424311000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Diethyldithiocarbamic acid, sodium salt trihydrate, ACS reagent, Thermo Scientific Chemicals

Catalog number: 424311000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 424311000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
20624-25-3
IUPAC Name
sodium (diethylcarbamothioyl)sulfanide trihydrate
Molecular Formula
C5H16NNaO3S2
InChI Key
WWGXHTXOZKVJDN-UHFFFAOYSA-M
SMILES
O.O.O.[Na+].CCN(CC)C([S-])=S
Appearance (Color)
Off-white to light grey
Appearance (Form)
Powder or crystals
Infrared spectrum
Conforms
Sodium (Na)
30.5 to 32.5 % (as Na2SO4)
Solubility
(in water) Passes test

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text