Thermo Scientific Chemicals

Sodium hydroxide, 97+%, ACS reagent, pellets, Thermo Scientific Chemicals

Catalog number: 424330025
2.5 kg, Each
Thermo Scientific Chemicals

Sodium hydroxide, 97+%, ACS reagent, pellets, Thermo Scientific Chemicals

Catalog number: 424330025
2.5 kg, Each
Quantity
Catalog number: 424330025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1310-73-2
IUPAC Name
sodium hydroxide
Molecular Formula
HNaO
InChI Key
HEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES
[OH-].[Na+]
Appearance (Form)
Pellets
Titration with HCl
>=97.0 %
Heavy metals (as Ag)
=<0.002 %
Sodium carbonate (Na2CO3)
=<1.0 %
Nitrogen compounds (as N)
=<0.001 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text