Sodium phosphate, dibasic, 99+%, ACS reagent, suitable for buffer solutions
Sodium phosphate, dibasic, 99+%, ACS reagent, suitable for buffer solutions
Sodium phosphate, dibasic, 99+%, ACS reagent, suitable for buffer solutions
Sodium phosphate, dibasic, 99+%, ACS reagent, suitable for buffer solutions
Thermo Scientific Chemicals

Sodium phosphate, dibasic, 99+%, ACS reagent, suitable for buffer solutions

CAS: 7558-79-4 | HNa2O4P | 141.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number 424375000
Price (USD)
119.00
Each
-
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
119.00
Each
Add to cart
Chemical Identifiers
CAS7558-79-4
IUPAC Namedisodium hydrogen phosphate
Molecular FormulaHNa2O4P
InChI KeyBNIILDVGGAEEIG-UHFFFAOYSA-L
SMILES[Na+].[Na+].OP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder or granular powder
Titration excess HCl>=99.0 %
Loss on drying=<0.2 % (105°C)
Heavy metals (as Pb)=<10 ppm
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
pH buffer
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link