1x1 image pixel for data collection
Sodium phosphate, monobasic monohydrate, 98+%, ACS reagent
Sodium phosphate, monobasic monohydrate, 98+%, ACS reagent
Sodium phosphate, monobasic monohydrate, 98+%, ACS reagent
Sodium phosphate, monobasic monohydrate, 98+%, ACS reagent
Thermo Scientific Chemicals

Sodium phosphate, monobasic monohydrate, 98+%, ACS reagent

CAS: 10049-21-5 | H5NaO5P | 139.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
2.5 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number 424395000
Price (USD)
141.00
Each
In stock
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
141.00
Each
Add to cart
Chemical Identifiers
CAS10049-21-5
IUPAC Namesodium phosphoric acid hydrate
Molecular FormulaH5NaO5P
InChI KeyBBMHARZCALWXSL-UHFFFAOYSA-N
SMILESO.[Na+].OP(O)(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH98.0 to 102.0 %
Appearance (Color)Colorless to white
Appearance (Form)Crystalline powder or crystals
Heavy metals (ICP-OES)=<0.001 %
pH4.1 to 4.5 (5 % solution at 25.0°C)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection