Thermo Scientific Chemicals

Sodium sulfate, 99+%, ACS reagent, anhydrous, Thermo Scientific Chemicals

Catalog number: 424410010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Sodium sulfate, 99+%, ACS reagent, anhydrous, Thermo Scientific Chemicals

Catalog number: 424410010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 424410010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7757-82-6
IUPAC Name
disodium sulfate
Molecular Formula
Na2O4S
InChI Key
PMZURENOXWZQFD-UHFFFAOYSA-L
SMILES
[Na+].[Na+].[O-]S([O-])(=O)=O
Appearance (Color)
White
Appearance (Form)
Crystalline or granular powder
Potassium (K)
=<100 ppm
Titration after Ion exchange
>=99.0 %
Heavy metals (as Pb)
=<5 ppm (by ICP-OES)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text