Sodium sulfate, 99+%, ACS reagent, anhydrous, Thermo Scientific Chemicals
Sodium sulfate, 99+%, ACS reagent, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium sulfate, 99+%, ACS reagent, anhydrous, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 kg
1 kg
2.5 kg
Packaging:
View all
Plastic drum
Catalog number 424410100
Price (USD)/ Each
419.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
494.00 
Save 74.35 (15%)
-
Add to cart
Quantity:
10 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)/ Each
419.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
494.00 
Save 74.35 (15%)
Add to cart
Sodium sulfate, 99+%, ACS reagent, anhydrous, Thermo Scientific Chemicals
Catalog number424410100
Price (USD)/ Each
419.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
494.00 
Save 74.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namedisodium sulfate
Molecular FormulaNa2O4S
InChI KeyPMZURENOXWZQFD-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]S([O-])(=O)=O
Molecular Weight (g/mol)142.04
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline or granular powder
Potassium (K)=<100 ppm
Titration after Ion exchange>=99.0 %
Heavy metals (as Pb)=<5 ppm (by ICP-OES)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only