Thermo Scientific Chemicals

Sodium sulfide nonahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 424420250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Sodium sulfide nonahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 424420250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 424420250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1313-84-4
IUPAC Name
disodium nonahydrate sulfanediide
Molecular Formula
H18Na2O9S
InChI Key
ZGHLCBJZQLNUAZ-UHFFFAOYSA-N
SMILES
O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--]
Appearance (Form)
Adhering crystals and/or chunks
Iron (Fe)
Passes test
Ammonium (NH4)
=<50 ppm
Appearance (Color)
Colorless to light yellow
Titration Iodimetric
>=98.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text