Thermo Scientific Chemicals

L+-Tartaric Acid, Disodium Salt, Dihydrate, Reagent ACS (crystals), ≥99%, Thermo Scientific Chemicals

Catalog number: 424440025
2.5 kg, Each
Thermo Scientific Chemicals

L+-Tartaric Acid, Disodium Salt, Dihydrate, Reagent ACS (crystals), ≥99%, Thermo Scientific Chemicals

Catalog number: 424440025
2.5 kg, Each
Quantity
Catalog number: 424440025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
6106-24-7
IUPAC Name
disodium (2R,3R)-2,3-dihydroxybutanedioate dihydrate
Molecular Formula
C4H8Na2O8
InChI Key
FGJLAJMGHXGFDE-UDMGOBQRNA-L
SMILES
O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O
Appearance (Color)
White
Infrared spectrum
Conforms
Heavy metals (as Pb)
=<5 ppm
Ammonium (NH4)
=<30 ppm
Iron (Fe)
=<10 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text