Thermo Scientific Chemicals

Tetraphenylboron sodium, ACS reagent, Thermo Scientific Chemicals

Catalog number: 424455000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Tetraphenylboron sodium, ACS reagent, Thermo Scientific Chemicals

Catalog number: 424455000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 424455000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
143-66-8
IUPAC Name
sodium tetraphenylboranuide
Molecular Formula
C24H20BNa
InChI Key
HFSRCEJMTLMDLI-UHFFFAOYSA-N
SMILES
[Na+].C1=CC=C(C=C1)[B-](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Infrared spectrum
Conforms
Appearance (Form)
Powder or crystals
Loss on drying
=<0.5 % (105°C)
Appearance (Color)
White to light beige
Gravimetry
>=99.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text