D(+)-Sucrose, ACS reagent, Thermo Scientific Chemicals
D(+)-Sucrose, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

D(+)-Sucrose, ACS reagent, Thermo Scientific Chemicals

D(+)-Sucrose, CAS # 57-50-1, is a disaccharide used in a variety of biochemical applications such as protein purification. It is also known as sugar, sucrose, alpha-D-glucopyranoside, and D-(+)-saccharose.
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Catalog number 424500010
Price (USD)/ Each
79.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
93.00 
Save 13.35 (14%)
-
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)/ Each
79.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
93.00 
Save 13.35 (14%)
Add to cart
D(+)-Sucrose, ACS reagent, Thermo Scientific Chemicals
Catalog number424500010
Price (USD)/ Each
79.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
93.00 
Save 13.35 (14%)
-
Add to cart
Chemical Identifiers
IUPAC Name(2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular FormulaC12H22O11
InChI KeyCZMRCDWAGMRECN-PWPRYFECNA-N
SMILESOC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
Molecular Weight (g/mol)342.30
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals
Iron (Fe)=<5 ppm
SO4 and SO3 (as SO4)=<0.005 %
Infrared spectrumConforms
Loss on drying=<0.03 % (105°C)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• D(+)-Sucrose is a disaccharide carbohydrate
• It is also known as sugar, sucrose, alpha-D-glucopyranoside, and D-(+)-saccharose
Applications
• D(+)-Sucrose can be used for protein and nucleic acid purification
• Its fatty acid esters have laboratory research applications in transdermal systems
• It is also used for sucrose-gradient centrifugation, which may be employed for the selective isolation of Nocardia species from soil
RUO – Research Use Only

General References:

  1. Jain, A.; Shore, A. M.; Jonnalagadda, S. C.; Ramanujachary, K. V.; Mugweru, A. Conversion of fructose, glucose, and sucrose to 5-hydroxymethyl-2-furfural over mesoporous zirconium phosphate catalyst. Appl. Catal. A 2015, 489, 72-76.
  2. Chen, Y.; Zhang, H.; Li, A.; Zhuo, K. Conductivities for ZnSO4/MgSO4+ sucrose/trehalose + water systems at 298.15 K. Fluid Phase Equilib. 2015, 388, 78-83.