Thermo Scientific Chemicals

5-Sulfosalicylic acid dihydrate, ACS reagent, Thermo Scientific Chemicals

Catalog number: 424515000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

5-Sulfosalicylic acid dihydrate, ACS reagent, Thermo Scientific Chemicals

Catalog number: 424515000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 424515000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5965-83-3
IUPAC Name
2-hydroxy-5-sulfobenzoic acid; bis(methane)
Molecular Formula
C9H14O6S
InChI Key
NFYHZVWMQHQKRU-UHFFFAOYSA-N
SMILES
C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O
Chloride (Cl)
=<0.001 %
Iron (Fe)
=<0.001 %
Heavy metals (as Pb)
=<0.002 %
Residue after ignition
=<0.1 %
Salicylic acid
=<0.04 % (HOC6H4COOH)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text