Thiourea, 99+%, ACS reagent, Thermo Scientific Chemicals
Thiourea, 99+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Thiourea, 99+%, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 424542500
Price (USD)/ Each
88.65
Online exclusive
104.00 
Save 15.35 (15%)
-
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
88.65
Online exclusive
104.00 
Save 15.35 (15%)
Add to cart
Thiourea, 99+%, ACS reagent, Thermo Scientific Chemicals
Catalog number424542500
Price (USD)/ Each
88.65
Online exclusive
104.00 
Save 15.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namethiourea
Molecular FormulaCH4N2S
InChI KeyUMGDCJDMYOKAJW-UHFFFAOYSA-N
SMILESNC(N)=S
Molecular Weight (g/mol)76.12
View more
SpecificationsSpecification SheetSpecification Sheet
Residue after ignition=<0.1 %
Solubility(16.5g in 100ml water,25°C) Passes test
Appearance (Color)White
Melting point174°C to 177°C
Titration Argentometric>=99.0 % (On dried basis)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only