Thermo Scientific Chemicals

Tris(hydroxymethyl)aminomethane, 99.8%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 424570025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Tris(hydroxymethyl)aminomethane, 99.8%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 424570025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 424570025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
77-86-1
IUPAC Name
2-amino-2-(hydroxymethyl)propane-1,3-diol
Molecular Formula
C4H11NO3
InChI Key
LENZDBCJOHFCAS-UHFFFAOYSA-N
SMILES
NC(CO)(CO)CO
Appearance (Color)
White
Appearance (Form)
Crystalline powder or crystals and/or chunks
Infrared spectrum
Conforms
Titration with HCl
99.8 to 100.1 % (On dry basis)
Water
=<2 % (K.F.)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text