Urea, 99%, Thermo Scientific Chemicals
Urea, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Urea, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number 424585000
Price (USD)/ Each
72.65
Online exclusive
80.30 
Save 7.65 (10%)
-
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
72.65
Online exclusive
80.30 
Save 7.65 (10%)
Add to cart
Urea, 99%, Thermo Scientific Chemicals
Catalog number424585000
Price (USD)/ Each
72.65
Online exclusive
80.30 
Save 7.65 (10%)
-
Add to cart
Chemical Identifiers
CAS57-13-6
IUPAC Nameurea
Molecular FormulaCH4N2O
InChI KeyXSQUKJJJFZCRTK-UHFFFAOYSA-N
SMILESNC(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point132°C to 135°C
Appearance (Form)Crystals
Infrared spectrumConforms
Titration with HClO499.0 to 100.5 %
Heavy metals (as Pb)=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only