4-Methylsulphonyl benzaldehyde, 95%, Thermo Scientific Chemicals
4-Methylsulphonyl benzaldehyde, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Methylsulphonyl benzaldehyde, 95%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
2.5 g
Packaging:
Glass bottle
Catalog number 424900025
Price (USD)/ Each
138.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
163.00 
Save 24.35 (15%)
-
Add to cart
Quantity:
2.5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
138.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
163.00 
Save 24.35 (15%)
Add to cart
4-Methylsulphonyl benzaldehyde, 95%, Thermo Scientific Chemicals
Catalog number424900025
Price (USD)/ Each
138.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
163.00 
Save 24.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name4-methanesulfonylbenzaldehyde
Molecular FormulaC8H8O3S
InChI KeyPSVPUHBSBYJSMQ-UHFFFAOYSA-N
SMILESCS(=O)(=O)C1=CC=C(C=O)C=C1
Molecular Weight (g/mol)184.21
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to beige to light brown
GC>=94 %
Appearance (Form)Crystalline powder or crystals
Melting point157°C to 163°C
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only