Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Use standard filter papers with Thermo Scientific™ Nalgene™ Polypropylene Analytical Funnels. Featuring long stems and external ribbing that eliminates air lock, these funnels are a shatterproof alternative to glass and chemically resistant to a wide variety of lab chemicals and reagents. These funnels make a valuble contribution to lab safety programs.
Diameter (Metric)Top | 158 mm |
Capacity (Metric) | 976 mL |
Diameter (Metric) Stem | 13 mm |
Material | Polypropylene |
Autoclavable | Yes |
Description | Funnel Analytical PP 160mm |
Length (Metric) Stem | 152 mm |
Quantity | 2/Pk.; 12/Cs. |
Unit Size | Pack of 2 |
Diameter (Metric)Top | 35 mm |
Capacity (Metric) | 10 mL |
Diameter (Metric) Stem | 6 mm |
Material | Polypropylene |
Autoclavable | Yes |
Description | Funnel Analytical PP 35mm |
Length (Metric) Stem | 50 mm |
Quantity | 12/Pk.; 36/Cs. |
Unit Size | Case of 36 |
Diameter (Metric)Top | 35 mm |
Capacity (Metric) | 10 mL |
Diameter (Metric) Stem | 6 mm |
Material | Polypropylene |
Autoclavable | Yes |
Description | Funnel Analytical PP 35mm |
Length (Metric) Stem | 50 mm |
Quantity | 12/Pk.; 36/Cs. |
Unit Size | Pack of 12 |
Diameter (Metric)Top | 54 mm |
Capacity (Metric) | 41 mL |
Diameter (Metric) Stem | 7 mm |
Material | Polypropylene |
Autoclavable | Yes |
Description | Funnel Analytical PP 55mm |
Length (Metric) Stem | 61 mm |
Quantity | 12/Pk.; 36/Cs. |
Unit Size | Case of 36 |
Diameter (Metric)Top | 54 mm |
Capacity (Metric) | 41 mL |
Diameter (Metric) Stem | 7 mm |
Material | Polypropylene |
Autoclavable | Yes |
Description | Funnel Analytical PP 55mm |
Length (Metric) Stem | 61 mm |
Quantity | 12/Pk.; 36/Cs. |
Unit Size | Pack of 12 |
Diameter (Metric)Top | 63 mm |
Capacity (Metric) | 64 mL |
Diameter (Metric) Stem | 7 mm |
Material | Polypropylene |
Autoclavable | Yes |
Description | Funnel Analytical PP 65mm |
Length (Metric) Stem | 58 mm |
Quantity | 12/Pk.; 36/Cs. |
Unit Size | Case of 36 |
Diameter (Metric)Top | 63 mm |
Capacity (Metric) | 64 mL |
Diameter (Metric) Stem | 7 mm |
Material | Polypropylene |
Autoclavable | Yes |
Description | Funnel Analytical PP 65mm |
Length (Metric) Stem | 58 mm |
Quantity | 12/Pk.; 36/Cs. |
Unit Size | Pack of 12 |
Diameter (Metric)Top | 76 mm |
Capacity (Metric) | 114 mL |
Diameter (Metric) Stem | 7 mm |
Material | Polypropylene |
Autoclavable | Yes |
Description | Funnel Analytical PP 75mm |
Length (Metric) Stem | 80 mm |
Quantity | 6/Pk.; 36/Cs. |
Unit Size | Case of 36 |
Diameter (Metric)Top | 76 mm |
Capacity (Metric) | 114 mL |
Diameter (Metric) Stem | 7 mm |
Material | Polypropylene |
Autoclavable | Yes |
Description | Funnel Analytical PP 75mm |
Length (Metric) Stem | 80 mm |
Quantity | 6/Pk.; 36/Cs. |
Unit Size | Pack of 6 |
Diameter (Metric)Top | 98 mm |
Capacity (Metric) | 254 mL |
Diameter (Metric) Stem | 9 mm |
Material | Polypropylene |
Autoclavable | Yes |
Description | Funnel Analytical PP 100mm |
Length (Metric) Stem | 100 mm |
Quantity | 4/Pk.; 24/Cs. |
Unit Size | Case of 24 |
Catalog Number | Specifications | Unit Size | Diameter (Metric)Top | Capacity (Metric) | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
4250-0160PK also known as 4250-0160 | Pack of 2 | 158 mm | 976 mL | 70.60 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
4250-0035 | Case of 36 | 35 mm | 10 mL | 119.70 Online price 171.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
4250-0035PK also known as 4250-0035 | Pack of 12 | 35 mm | 10 mL | 67.70 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
4250-0055 | Case of 36 | 54 mm | 41 mL | 186.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
4250-0055PK also known as 4250-0055 | Pack of 12 | 54 mm | 41 mL | 73.30 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
4250-0065 | Case of 36 | 63 mm | 64 mL | 154.00 Online price 220.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
4250-0065PK also known as 4250-0065 | Pack of 12 | 63 mm | 64 mL | 86.70 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
4250-0075 | Case of 36 | 76 mm | 114 mL | 280.00 Online price 312.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
4250-0075PK also known as 4250-0075 | Pack of 6 | 76 mm | 114 mL | 61.40 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
4250-0100 | Case of 24 | 98 mm | 254 mL | 296.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|