Thermo Scientific™
RadEye™ G Series Personal Dose Rate Meters
Catalog number: 4250674
Have Questions?
Thermo Scientific™
RadEye™ G Series Personal Dose Rate Meters
Catalog number: 4250674
Have Questions?
Continuously monitor your radiation exposure in any nuclear or radiological emergency. The Thermo Scientific™ RadEye™ G Series of personal radiation detectors provide first responders with quick, reliable gamma and dose rate measurements. These light-weight, handheld Geiger counters detect and measure very low gamma energies; even the smallest change in radiation rates are displayed immediately, while coincidentally occurring fluctuations are suppressed. For hazmat teams investigating potentially explosive atmospheres, intrinsically safe RadEye G models meet the latest ATEX standards.
Catalog Number
4250674
Unit Size
Each
Description
RadEye G with Black Label
Price (USD)
Full specifications
DescriptionRadEye G with Black Label
Dimensions (L x W x H)3.78 x 1.25 x 2.4 in. (9.6 x 3.1 x 6.1 cm)
Protection DegreeIP 65 according to EN 60529
Energy Range45 keV to 1.3 MeV (+/-30%)
Battery Type2 AAA
Product TypePersonal Dose Rate Meter
Measurement Range50 µR/h to 10 R/h
Detector TypeEnergy-compensated Geiger-Mueller tube
Overrange Threshold1000 R/h
Count RateFor Cs-137 (66 keV): 1.7 cps per μSv/h
Weight (Metric)0.18 kg
UnitsR and R/h
Battery Life900 hr.
Dose Rate Limit10 R/h
Weight (English)0.41 lb.
Dose Rangeup to 1000 R
Unit SizeEach
Catalog Number
Unit Size
Each
Description
RadEye G with Yellow Label
Price (USD)
Full specifications
DescriptionRadEye G with Yellow Label
Dimensions (L x W x H)3.78 x 1.25 x 2.4 in. (9.6 x 3.1 x 6.1 cm)
Protection DegreeIP 65 according to EN 60529
Energy Range45 keV to 1.3 MeV (+/-30%)
Battery Type2 AAA
Product TypePersonal Dose Rate Meter
Measurement Range50 µR/h to 10 R/h
Detector TypeEnergy-compensated Geiger-Mueller tube
Overrange Threshold1000 R/h
Count RateFor Cs-137 (66 keV): 1.7 cps per μSv/h
Weight (Metric)0.18 kg
UnitsR and R/h
Battery Life900 hr.
Dose Rate Limit10 R/h
Weight (English)0.41 lb.
Unit SizeEach
Catalog Number
Unit Size
Each
Description
RadEye G-10 with Red Label
Price (USD)
Full specifications
DescriptionRadEye G-10 with Red Label
Dimensions (L x W x H)3.78 x 1.25 x 2.4 in. (9.6 x 3.1 x 6.1 cm)
Protection DegreeIP 65 according to EN 60529
Energy Range50 keV to 3 MeV in accordance with IEC 60846-1 (incl. angular dependency)
Battery Type2 AAA
Product TypePersonal Dose Rate Meter
Measurement Range0.5 μSv/h to 100 mSv/h
Detector TypeEnergy-compensated Geiger-Mueller tube
Overrange Thresholdup to 10 Sv/h
Count RateFor Cs-137 (66 keV): 1.7 cps per μSv/h
Weight (Metric)0.18 kg
UnitsH*(10) in Sv and Sv/h
Battery Life900 hr.
Weight (English)0.41 lb.
Dose RangeH*(10) up to 10 Sv
Unit SizeEach
Catalog Number
Unit Size
Each
Description
RadEye G-10-Ex with Orange Label
Price (USD)
Full specifications
ATEX ClassificationII 2G Ex ia IIB T4 IBExU10ATEX1096
DescriptionRadEye G-10-Ex with Orange Label
Dimensions (L x W x H)3.78 x 1.25 x 2.4 in. (9.6 x 3.1 x 6.1 cm)
Protection DegreeIP 65 according to EN 60529
Energy Range50 keV to 3 MeV in accordance with IEC 60846-1 (incl. angular dependency)
Battery Type2 AAA
Product TypePersonal Radiation Detector
Measurement Range0.5 µSv/h to 100 mSv/h [50 urem/h - 10 rem/h]
Detector TypeEnergy-compensated Geiger-Mueller tube
Overrange Thresholdup to 10 Sv/h
Count RateFor Cs-137 (622 keV): 1.7 cps per μSv/h
Weight (Metric)0.18 kg
UnitsH*(10) in Sv and Sv/h
Battery Life900 hr.
Weight (English)0.41 lb.
Dose RangeH*(10) up to 10 Sv
Unit SizeEach
Catalog Number
Unit Size
Each
Description
RadEye G-Ex with Orange Label
Price (USD)
Full specifications
ATEX ClassificationII 2G Ex ia IIB T4 IBExU10ATEX1096
DescriptionRadEye G-Ex with Orange Label
Dimensions (L x W x H)3.78 x 1.25 x 2.4 in. (9.6 x 3.1 x 6.1 cm)
Protection DegreeIP 65 according to EN 60529
Energy Range45 keV to 1.3 MeV (+/-30%)
Battery Type2 AAA
Product TypePersonal Radiation Detector
Measurement Range50µR/h to 10R/h
Detector TypeEnergy-compensated Geiger-Mueller tube
Overrange Threshold1000 R/h
Count RateFor Cs-137 (622 keV): 17 cps per mR/h
Weight (Metric)0.18 kg
UnitsR and R/h
Battery Life900 hr.
Weight (English)0.41 lb.
Dose Rangeup to 1000 R
Unit SizeEach
Catalog Number
Unit Size
Each
Description
RadEye GF-Ex with Orange Label
Price (USD)
Full specifications
ATEX ClassificationII 2G Ex ia IIB T4 IBExU10ATEX1096
DescriptionRadEye GF-Ex with Orange Label
Dimensions (L x W x H)3.78 x 1.25 x 2.4 in. (9.6 x 3.1 x 6.1 cm)
Protection DegreeIP 65 according to EN 60529
Energy Range45 keV to 1.3 MeV (+/-30%)
Battery Type2 AAA
Product TypePersonal Radiation Detector
Measurement Range500 μR/h to 300 R/h
Detector TypeEnergy-compensated Geiger-Mueller tube
Overrange Threshold1000 R/h
Count RateFor Cs-137 (622 keV): 1.3 cps per mR/h
Weight (Metric)0.18 kg
UnitsR and R/h
Battery Life900 hr.
Weight (English)0.41 lb.
Dose Rangeup to 1000 R
Unit SizeEach
Catalog Number
Unit Size
Each
Description
RadEye GF-10-Ex with Orange Label
Price (USD)
Full specifications
ATEX ClassificationII 2G Ex ia IIB T4 IBExU10ATEX1096
SensitivityCs-137, 660 keV, ˜0.13cps/μSv/h
DescriptionRadEye GF-10-Ex with Orange Label
Dimensions (L x W x H)3.78 x 1.25 x 2.4 in. (9.6 x 3.1 x 6.1 cm)
Protection DegreeIP 65 according to EN 60529
Energy Range50 keV to 3 MeV in accordance with IEC 60846-1 (incl. angular dependency)
Battery Type2 AAA
Product TypePersonal Radiation Detector
Measurement Range5 µSv/h to 3 Sv/h [500 urem/h - 300 rem/h]
Detector TypeEnergy-compensated Geiger-Mueller tube
Overrange Thresholdup to 10 Sv/h
Count RateFor Cs-137 (622 keV): 0.13 cps per μSv/h
Weight (Metric)0.18 kg
UnitsH*(10) in Sv and Sv/h
Battery Life900 hr.
Weight (English)0.41 lb.
Dose RangeH*(10) up to 10 Sv
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Radeye GF with Yellow Label
Price (USD)
Full specifications
DescriptionRadeye GF with Yellow Label
Dimensions (L x W x H)3.78 x 1.25 x 2.4 in. (9.6 x 3.1 x 6.1 cm)
Protection DegreeIP 65 according to EN 60529
Energy Range45 keV to 1.3 MeV (+/-30%)
Battery Type2 AAA
Product TypePersonal Radiation Detector
Measurement Range500 μR/h - 300 R/h
Detector TypeEnergy-compensated Geiger-Mueller tube
Overrange Threshold1000 R/h
Count RateFor Cs-137 (662 keV): 1.3 cps per mR/h
Weight (Metric)0.18 kg
UnitsR and R/h
Battery Life900 hr.
Weight (English)0.41 lb.
Dose Rangeup to 1000 R
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Radeye GF-10 with Yellow Label
Price (USD)
Full specifications
DescriptionRadeye GF-10 with Yellow Label
Dimensions (L x W x H)3.78 x 1.25 x 2.4 in. (9.6 x 3.1 x 6.1 cm)
Protection DegreeIP 65 according to EN 60529
Energy Range50 keV to 3 MeV in accordance with IEC 60846-1 (incl. angular dependency)
Battery Type2 AAA
Product TypePersonal Radiation Detector
Measurement Range5 µSv/h to 3 Sv/h [500 urem/h - 300 rem/h]
Detector TypeEnergy-compensated Geiger-Mueller tube
Overrange Thresholdup to 10 Sv/h
Count RateFor Cs-137 (662 keV): 0.13 cps per μSv/h
Weight (Metric)0.18 kg
UnitsH*(10) in Sv and Sv/h
Battery Life900 hr.
Weight (English)0.41 lb.
Dose RangeH*(10) up to 10 Sv
Unit SizeEach
Showing 9 of 9
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)
4250674Full specifications
EachRadEye G with Black LabelRequest A Quote
DescriptionRadEye G with Black Label
Dimensions (L x W x H)3.78 x 1.25 x 2.4 in. (9.6 x 3.1 x 6.1 cm)
Protection DegreeIP 65 according to EN 60529
Energy Range45 keV to 1.3 MeV (+/-30%)
Battery Type2 AAA
Product TypePersonal Dose Rate Meter
Measurement Range50 µR/h to 10 R/h
Detector TypeEnergy-compensated Geiger-Mueller tube
Overrange Threshold1000 R/h
Count RateFor Cs-137 (66 keV): 1.7 cps per μSv/h
Weight (Metric)0.18 kg
UnitsR and R/h
Battery Life900 hr.
Dose Rate Limit10 R/h
Weight (English)0.41 lb.
Dose Rangeup to 1000 R
Unit SizeEach
425067401Full specifications
EachRadEye G with Yellow LabelRequest A Quote
DescriptionRadEye G with Yellow Label
Dimensions (L x W x H)3.78 x 1.25 x 2.4 in. (9.6 x 3.1 x 6.1 cm)
Protection DegreeIP 65 according to EN 60529
Energy Range45 keV to 1.3 MeV (+/-30%)
Battery Type2 AAA
Product TypePersonal Dose Rate Meter
Measurement Range50 µR/h to 10 R/h
Detector TypeEnergy-compensated Geiger-Mueller tube
Overrange Threshold1000 R/h
Count RateFor Cs-137 (66 keV): 1.7 cps per μSv/h
Weight (Metric)0.18 kg
UnitsR and R/h
Battery Life900 hr.
Dose Rate Limit10 R/h
Weight (English)0.41 lb.
Unit SizeEach
4250676Full specifications
EachRadEye G-10 with Red LabelRequest A Quote
DescriptionRadEye G-10 with Red Label
Dimensions (L x W x H)3.78 x 1.25 x 2.4 in. (9.6 x 3.1 x 6.1 cm)
Protection DegreeIP 65 according to EN 60529
Energy Range50 keV to 3 MeV in accordance with IEC 60846-1 (incl. angular dependency)
Battery Type2 AAA
Product TypePersonal Dose Rate Meter
Measurement Range0.5 μSv/h to 100 mSv/h
Detector TypeEnergy-compensated Geiger-Mueller tube
Overrange Thresholdup to 10 Sv/h
Count RateFor Cs-137 (66 keV): 1.7 cps per μSv/h
Weight (Metric)0.18 kg
UnitsH*(10) in Sv and Sv/h
Battery Life900 hr.
Weight (English)0.41 lb.
Dose RangeH*(10) up to 10 Sv
Unit SizeEach
425067660Full specifications
EachRadEye G-10-Ex with Orange LabelRequest A Quote
ATEX ClassificationII 2G Ex ia IIB T4 IBExU10ATEX1096
DescriptionRadEye G-10-Ex with Orange Label
Dimensions (L x W x H)3.78 x 1.25 x 2.4 in. (9.6 x 3.1 x 6.1 cm)
Protection DegreeIP 65 according to EN 60529
Energy Range50 keV to 3 MeV in accordance with IEC 60846-1 (incl. angular dependency)
Battery Type2 AAA
Product TypePersonal Radiation Detector
Measurement Range0.5 µSv/h to 100 mSv/h [50 urem/h - 10 rem/h]
Detector TypeEnergy-compensated Geiger-Mueller tube
Overrange Thresholdup to 10 Sv/h
Count RateFor Cs-137 (622 keV): 1.7 cps per μSv/h
Weight (Metric)0.18 kg
UnitsH*(10) in Sv and Sv/h
Battery Life900 hr.
Weight (English)0.41 lb.
Dose RangeH*(10) up to 10 Sv
Unit SizeEach
425067460Full specifications
EachRadEye G-Ex with Orange LabelRequest A Quote
ATEX ClassificationII 2G Ex ia IIB T4 IBExU10ATEX1096
DescriptionRadEye G-Ex with Orange Label
Dimensions (L x W x H)3.78 x 1.25 x 2.4 in. (9.6 x 3.1 x 6.1 cm)
Protection DegreeIP 65 according to EN 60529
Energy Range45 keV to 1.3 MeV (+/-30%)
Battery Type2 AAA
Product TypePersonal Radiation Detector
Measurement Range50µR/h to 10R/h
Detector TypeEnergy-compensated Geiger-Mueller tube
Overrange Threshold1000 R/h
Count RateFor Cs-137 (622 keV): 17 cps per mR/h
Weight (Metric)0.18 kg
UnitsR and R/h
Battery Life900 hr.
Weight (English)0.41 lb.
Dose Rangeup to 1000 R
Unit SizeEach
425067470Full specifications
EachRadEye GF-Ex with Orange LabelRequest A Quote
ATEX ClassificationII 2G Ex ia IIB T4 IBExU10ATEX1096
DescriptionRadEye GF-Ex with Orange Label
Dimensions (L x W x H)3.78 x 1.25 x 2.4 in. (9.6 x 3.1 x 6.1 cm)
Protection DegreeIP 65 according to EN 60529
Energy Range45 keV to 1.3 MeV (+/-30%)
Battery Type2 AAA
Product TypePersonal Radiation Detector
Measurement Range500 μR/h to 300 R/h
Detector TypeEnergy-compensated Geiger-Mueller tube
Overrange Threshold1000 R/h
Count RateFor Cs-137 (622 keV): 1.3 cps per mR/h
Weight (Metric)0.18 kg
UnitsR and R/h
Battery Life900 hr.
Weight (English)0.41 lb.
Dose Rangeup to 1000 R
Unit SizeEach
425067670Full specifications
EachRadEye GF-10-Ex with Orange LabelRequest A Quote
ATEX ClassificationII 2G Ex ia IIB T4 IBExU10ATEX1096
SensitivityCs-137, 660 keV, ˜0.13cps/μSv/h
DescriptionRadEye GF-10-Ex with Orange Label
Dimensions (L x W x H)3.78 x 1.25 x 2.4 in. (9.6 x 3.1 x 6.1 cm)
Protection DegreeIP 65 according to EN 60529
Energy Range50 keV to 3 MeV in accordance with IEC 60846-1 (incl. angular dependency)
Battery Type2 AAA
Product TypePersonal Radiation Detector
Measurement Range5 µSv/h to 3 Sv/h [500 urem/h - 300 rem/h]
Detector TypeEnergy-compensated Geiger-Mueller tube
Overrange Thresholdup to 10 Sv/h
Count RateFor Cs-137 (622 keV): 0.13 cps per μSv/h
Weight (Metric)0.18 kg
UnitsH*(10) in Sv and Sv/h
Battery Life900 hr.
Weight (English)0.41 lb.
Dose RangeH*(10) up to 10 Sv
Unit SizeEach
425067475Full specifications
EachRadeye GF with Yellow LabelRequest A Quote
DescriptionRadeye GF with Yellow Label
Dimensions (L x W x H)3.78 x 1.25 x 2.4 in. (9.6 x 3.1 x 6.1 cm)
Protection DegreeIP 65 according to EN 60529
Energy Range45 keV to 1.3 MeV (+/-30%)
Battery Type2 AAA
Product TypePersonal Radiation Detector
Measurement Range500 μR/h - 300 R/h
Detector TypeEnergy-compensated Geiger-Mueller tube
Overrange Threshold1000 R/h
Count RateFor Cs-137 (662 keV): 1.3 cps per mR/h
Weight (Metric)0.18 kg
UnitsR and R/h
Battery Life900 hr.
Weight (English)0.41 lb.
Dose Rangeup to 1000 R
Unit SizeEach
425067675Full specifications
EachRadeye GF-10 with Yellow LabelRequest A Quote
DescriptionRadeye GF-10 with Yellow Label
Dimensions (L x W x H)3.78 x 1.25 x 2.4 in. (9.6 x 3.1 x 6.1 cm)
Protection DegreeIP 65 according to EN 60529
Energy Range50 keV to 3 MeV in accordance with IEC 60846-1 (incl. angular dependency)
Battery Type2 AAA
Product TypePersonal Radiation Detector
Measurement Range5 µSv/h to 3 Sv/h [500 urem/h - 300 rem/h]
Detector TypeEnergy-compensated Geiger-Mueller tube
Overrange Thresholdup to 10 Sv/h
Count RateFor Cs-137 (662 keV): 0.13 cps per μSv/h
Weight (Metric)0.18 kg
UnitsH*(10) in Sv and Sv/h
Battery Life900 hr.
Weight (English)0.41 lb.
Dose RangeH*(10) up to 10 Sv
Unit SizeEach
Showing 9 of 9
  • Menu driven interface can be optimized to application
  • Large, clear, backlit display for error free readings
  • High range versions for high turn back levels up to 300 R/h (3 Sv/h)
  • Intrinsically safe versions for potentially explosive environments
  • Large energy compensated GM tube for precise dose rate measurement of gamma and X-ray
  • High sensitivity to low energy gamma radiation in harsh environments
  • Durable – shock resistant design
  • 900 hour operation with 2 AAA batteries
  • Bright LED allows for operation in smoke and darkness
  • One hot and four advanced buttons – easy to use Alarm relay output – for area monitor application
  • Designed to exceed ANSI N42.33 standard
  • Intrinsically safe , ATEX certified
  • Data analysis can be done by an optional Windows™-based PC-software and an accompanying reader device
View our online training here.