1x1 image pixel for data collection
RadEye PRD4 and PRD-ER4 Personal Radiation Detector
RadEye PRD4 and PRD-ER4 Personal Radiation Detector
Actual product may vary
RadEye PRD4 and PRD-ER4 Personal Radiation Detector
RadEye PRD4 and PRD-ER4 Personal Radiation Detector
Thermo Scientific™
RadEye PRD4 and PRD-ER4 Personal Radiation Detector
The Thermo Scientific™ RadEye™ PRD4 and PRD-ER4 Personal Radiation Detectors provide highly sensitive radiation detection capability with little to no nuisance alarms.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
425069162RadEye PRD4-KIT, contains 1 ea. RadEye PRD4, Lutetium Test Adapter, Bluetooth adapter, Desktop holder + USB connection cable, Software package RadEye.exe, Holster and spare AAA batteries packaged in a hardened plastic case.
425067126RadEye PRD4, Pocket–sized personal radiation detector
425067127RadEye PRD-ER4, Pocket–sized Personal Radiation Detector
425069160RadEye PRD4 Charger package contains 1ea. RadEye PRD4, desktop charger, charging back, and batteries
425069161RadEye PRD4 Charger/BTLE package contains 1ea. RadEye PRD4, desktop charger, BTLE charging back, and batteries
425069163RadEye PRD4 Vehicle Kit, contains 1 ea. RadEye PRD4, Lutetium Test Adapter, Vehicle charging kit, Bluetooth adapter, Software package RadEye.exe, Holster and spare AAA rechargeable batteries
425069170RadEye PRD-ER4 Charger package contains 1ea. RadEye PRD-ER4, desktop charger, charging back, and batteries
425069171RadEye PRD-ER4 Charger/BTLE package contains 1ea. RadEye PRD-ER4, desktop charger, BTLE charging back, and batteries
425069172RadEye PRD-ER4-KIT, contains 1 ea. RadEye PRD-ER4, Lutetium Test Adapter, Bluetooth adapter, Desktop holder + USB connection cable, Software package RadEye.exe, Holster and spare AAA batteries packaged in a hardened plastic case.
425069173RadEye PRD-ER4 Vehicle Kit, contains 1 ea. RadEye PRD-ER4, Lutetium Test Adapter, Vehicle charging kit, Bluetooth adapter, Software package RadEye.exe, Holster and spare AAA rechargeable batteries
Catalog number 425069162
Price (USD)
-
Request A Quote
Description:
RadEye PRD4-KIT, contains 1 ea. RadEye PRD4, Lutetium Test Adapter, Bluetooth adapter, Desktop holder + USB connection cable, Software package RadEye.exe, Holster and spare AAA batteries packaged in a hardened plastic case.
Achieve highly sensitive radiation detection with few nuisance alarms with Thermo Scientific™ RadEye™ PRD4 and PRD-ER4 Personal Radiation Detectors. RadEye PRD4 and PRD-ER4 radiation detectors save users valuable time adjudicating alarms while increasing scanning throughput. Using Natural Background Rejection (NBR) Technology, the RadEye PRD4 and PRD-ER4 radiation detectors ignore fluctuations in naturally occurring radiation (NORM) while analyzing the energy imbalance of artificial radiation. Additionally, these radiation detectors identify the discovered material in a manner configurable for your operation or user skills.
The RadEye PRD4 and PRD-ER4 Personal Radiation Detectors enable accurate dose rate measurements across the broader gamma spectrum, providing a greater assurance of personal safety. The neutron indication capability of the RadEye PRD4 provides an additional layer of detection critical to determining the level of radiation threat.
Efficiency in the field
  • Larger display
  • Neutron indication capability
  • 25% improved sensitivity over previous model
  • Linear high dose rate detector
Easy information, smarter decisions
  • No retraining or relearning for infrequent users
  • Get results automatically, without the need to press buttons
  • Quickly guides you through next steps after an alarm
  • Simple 4 button design
  • Comprehensive data neatly organized and presented on screen
Rugged and practical
  • Can be worn in holster or standard service belt
  • Small and lightweight
  • Long battery life
  • Drop resistant to 1.5m
  • IP65
  • Can be operated in extreme temperatures
View our online training here.
Specifications
DescriptionRadEye PRD4-KIT, contains 1 ea. RadEye PRD4, Lutetium Test Adapter, Bluetooth adapter, Desktop holder + USB connection cable, Software package RadEye.exe, Holster and spare AAA batteries packaged in a hardened plastic case.
Unit SizeEach
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link
1x1 image pixel for data collection