Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Transfer powders with Thermo Scientific™ Nalgene™ Polypropylene Powder Funnels. Parallel stem minimizes bridging of powder and external ribbing prevents air locks. This shatter-proof alternative to glass is chemical resistant to a wide range of lab chemicals and reagents and can make a valuble contribution to lab safety programs.
Diameter (Metric)Top | 79mm |
Capacity (Metric) | 115 mL |
Diameter (Metric) Stem | 18 mm |
Material | Polypropylene |
Height (Metric) | 85 mm |
Autoclavable | Yes |
Description | Funnel Powder PP 80mm |
Length (Metric) Stem | 30 mm |
Quantity | 12/Pk.; 36/Cs. |
Unit Size | Pack of 12 |
Diameter (Metric)Top | 64 mm |
Capacity (Metric) | 68 mL |
Diameter (Metric) Stem | 17 mm |
Material | Polypropylene |
Height (Metric) | 67 mm |
Autoclavable | Yes |
Description | Funnel Powder PP 65mm |
Length (Metric) Stem | 22mm |
Quantity | 12/Pk.; 36/Cs. |
Unit Size | Case of 36 |
Diameter (Metric)Top | 64 mm |
Capacity (Metric) | 68 mL |
Diameter (Metric) Stem | 17 mm |
Material | Polypropylene |
Height (Metric) | 67 mm |
Autoclavable | Yes |
Description | Funnel Powder PP 65mm |
Length (Metric) Stem | 22mm |
Quantity | 12/Pk.; 36/Cs. |
Unit Size | Pack of 12 |
Diameter (Metric)Top | 79mm |
Capacity (Metric) | 115 mL |
Diameter (Metric) Stem | 18 mm |
Material | Polypropylene |
Height (Metric) | 85 mm |
Autoclavable | Yes |
Description | Funnel Powder PP 80mm |
Length (Metric) Stem | 30 mm |
Quantity | 12/Pk.; 36/Cs. |
Unit Size | Case of 36 |
Diameter (Metric)Top | 100 mm |
Capacity (Metric) | 243 mL |
Diameter (Metric) Stem | 23 mm |
Material | Polypropylene |
Height (Metric) | 105 mm |
Autoclavable | Yes |
Description | Funnel Powder PP 100mm |
Length (Metric) Stem | 34 mm |
Quantity | 6/Pk.; 24/Cs. |
Unit Size | Case of 24 |
Diameter (Metric)Top | 100 mm |
Capacity (Metric) | 243 mL |
Diameter (Metric) Stem | 23 mm |
Material | Polypropylene |
Height (Metric) | 105 mm |
Autoclavable | Yes |
Description | Funnel Powder PP 100mm |
Length (Metric) Stem | 34 mm |
Quantity | 6/Pk.; 24/Cs. |
Unit Size | Pack of 6 |
Diameter (Metric)Top | 145 mm |
Capacity (Metric) | 716 mL |
Diameter (Metric) Stem | 29 mm |
Material | Polypropylene |
Height (Metric) | 137 mm |
Autoclavable | Yes |
Description | Funnel Powder PP 150mm |
Length (Metric) Stem | 32 mm |
Quantity | 4/Pk.; 24/Cs. |
Unit Size | Case of 24 |
Diameter (Metric)Top | 145 mm |
Capacity (Metric) | 716 mL |
Diameter (Metric) Stem | 29 mm |
Material | Polypropylene |
Height (Metric) | 137 mm |
Autoclavable | Yes |
Description | Funnel Powder PP 150mm |
Length (Metric) Stem | 32 mm |
Quantity | 4/Pk.; 24/Cs. |
Unit Size | Pack of 4 |
Catalog Number | Specifications | Unit Size | Diameter (Metric)Top | Capacity (Metric) | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
4252-0080PK also known as 4252-0080 | Pack of 12 | 79mm | 115 mL | 98.10 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
4252-0065 | Case of 36 | 64 mm | 68 mL | 179.76 Online price 214.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
4252-0065PK also known as 4252-0065 | Pack of 12 | 64 mm | 68 mL | 84.20 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
4252-0080 | Case of 36 | 79mm | 115 mL | 175.00 Online price 250.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
4252-0100 | Case of 24 | 100 mm | 243 mL | 161.04 Online price 214.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
4252-0100PK also known as 4252-0100 | Pack of 6 | 100 mm | 243 mL | 63.20 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
4252-0150 | Case of 24 | 145 mm | 716 mL | 236.25 Online price 270.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
4252-0150PK also known as 4252-0150 | Pack of 4 | 145 mm | 716 mL | 53.10 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|