Thermo Scientific Chemicals

Ethyl picolinoylacetate, 95%, Thermo Scientific Chemicals

Catalog number: 425470025
2.5 g, Glass bottle, Each
Thermo Scientific Chemicals

Ethyl picolinoylacetate, 95%, Thermo Scientific Chemicals

Catalog number: 425470025
2.5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 425470025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
26510-52-1
Appearance (Color)
Clear straw yellow to brown
Appearance (Form)
Liquid
Infrared spectrum
Conforms
HPLC
>=94 %
Refractive index
1.5160 to 1.5190 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text