Thermo Scientific Chemicals

Methyl nicotinoylacetate, 95%, Thermo Scientific Chemicals

Catalog number: 425490025
2.5 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyl nicotinoylacetate, 95%, Thermo Scientific Chemicals

Catalog number: 425490025
2.5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 425490025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
54950-20-8
IUPAC Name
methyl 3-oxo-3-(pyridin-3-yl)propanoate
Molecular Formula
C9H9NO3
InChI Key
JUQKVXRLRKKRPL-UHFFFAOYSA-N
SMILES
COC(=O)CC(=O)C1=CC=CN=C1
Appearance (Color)
White to yellow to brown
Appearance (Form)
Crystalline powder or crystals and/or chunks
Infrared spectrum
Conforms
Melting point
34°C to 42°C
HPLC
>=94 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text