Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Catalog Number | Diameter (Metric)Top | Capacity (Metric) | Unit Size |
---|---|---|---|
4256-0234 | 62 mm | 62 mL | Case of 288 |
4256-0234PK also known as 4256-0234 | 62 mm | 62 mL | Pack of 12 |
4256-0414 | 98 mm | 270 mL | Case of 144 |
4256-0414PK also known as 4256-0414 | 98 mm | 270 mL | Pack of 12 |
4256-0638 | 150 mm | 1059 mL | Case of 72 |
4256-0638PK also known as 4256-0638 | 150 mm | 1059 mL | Pack of 12 |
Replace glass lab funnels with Thermo Scientific™ Nalgene™ Utility Funnels. These funnels are chemically resistant to a wide variety of lab chemicals and reagents and are shatterproof making them a valuble asset to lab safety programs.
Share catalog number, name or link