3,5-Dihydroxybenzaldehyde, 98%, Thermo Scientific Chemicals
3,5-Dihydroxybenzaldehyde, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,5-Dihydroxybenzaldehyde, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Packaging:
Glass bottle
Catalog number 425650050
Price (USD)/ Each
223.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
263.00 
Save 39.35 (15%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
223.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
263.00 
Save 39.35 (15%)
Add to cart
3,5-Dihydroxybenzaldehyde, 98%, Thermo Scientific Chemicals
Catalog number425650050
Price (USD)/ Each
223.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
263.00 
Save 39.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name3,5-dihydroxybenzaldehyde
Molecular FormulaC7H6O3
InChI KeyHAQLHRYUDBKTJG-UHFFFAOYSA-N
SMILESOC1=CC(C=O)=CC(O)=C1
Molecular Weight (g/mol)138.12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to grey to tan
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Loss on drying=<2 %
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only