1x1 image pixel for data collection
2-Benzyloxy-3-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
2-Benzyloxy-3-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
2-Benzyloxy-3-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
2-Benzyloxy-3-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Benzyloxy-3-methoxybenzaldehyde, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantityPackaging
4256700505 gGlass Bottle
Catalog number 425670050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS2011-06-5
IUPAC Name2-(benzyloxy)-3-methoxybenzaldehyde
Molecular FormulaC15H14O3
InChI KeyKXBGOVZWCRLLOR-UHFFFAOYSA-N
SMILESCOC1=CC=CC(C=O)=C1OCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to light yellow
Appearance (Form)Crystalline powder
Melting point44°C to 46°C
GC>=97.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Please enter sku to load default questions

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link