Thermo Scientific Chemicals

2,3-Diaminophenol, 97%, Thermo Scientific Chemicals

Catalog number: 425680010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

2,3-Diaminophenol, 97%, Thermo Scientific Chemicals

Catalog number: 425680010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 425680010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
59649-56-8
IUPAC Name
2,3-diaminophenol
Molecular Formula
C6H8N2O
InChI Key
PCAXITAPTVOLGL-UHFFFAOYSA-N
SMILES
NC1=C(N)C(O)=CC=C1
HPLC
>=96.0 %
Melting point
161°C to 165°C
Appearance (Color)
Light brown to dark brown
Appearance (Form)
Powder or crystalline powder
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text