cis-5-Norbornene-exo-2,3-dicarboxylic anhydride, 95%, Thermo Scientific Chemicals
cis-5-Norbornene-exo-2,3-dicarboxylic anhydride, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

cis-5-Norbornene-exo-2,3-dicarboxylic anhydride, 95%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Packaging:
Glass bottle
Catalog number 425740050
Price (USD)/ Each
162.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
191.00 
Save 28.35 (15%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
162.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
191.00 
Save 28.35 (15%)
Add to cart
cis-5-Norbornene-exo-2,3-dicarboxylic anhydride, 95%, Thermo Scientific Chemicals
Catalog number425740050
Price (USD)/ Each
162.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
191.00 
Save 28.35 (15%)
-
Add to cart
Chemical Identifiers
CAS2746-19-2
IUPAC Name(2R,6S)-4-oxatricyclo[5.2.1.0²,⁶]dec-8-ene-3,5-dione
Molecular FormulaC9H8O3
InChI KeyKNDQHSIWLOJIGP-CYDAGYADSA-N
SMILESO=C1OC(=O)[C@@H]2C3CC(C=C3)[C@H]12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to tan
Appearance (Form)Solid
Infrared spectrumConforms
Melting point140°C to 145°C
GC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only