Ethyl glyoxalate, technical, ∼50% in toluene, Thermo Scientific Chemicals
Ethyl glyoxalate, technical, ∼50% in toluene, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl glyoxalate, technical, ∼50% in toluene, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
Catalog number 425765000
Price (USD)/ Each
189.65
Online exclusive
211.00 
Save 21.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
189.65
Online exclusive
211.00 
Save 21.35 (10%)
Add to cart
Ethyl glyoxalate, technical, ∼50% in toluene, Thermo Scientific Chemicals
Catalog number425765000
Price (USD)/ Each
189.65
Online exclusive
211.00 
Save 21.35 (10%)
-
Add to cart
Chemical Identifiers
CAS924-44-7
IUPAC Nameethyl 2-oxoacetate
Molecular FormulaC4H6O3
InChI KeyDBPFRRFGLYGEJI-UHFFFAOYSA-N
SMILESCCOC(=O)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Solution
Titration excess HCl>=48 % (Aldehyde function)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only