Thermo Scientific Chemicals

4-Methyl-5-imidazolecarboxaldehyde, 99%, Thermo Scientific Chemicals

Catalog number: 425770250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Methyl-5-imidazolecarboxaldehyde, 99%, Thermo Scientific Chemicals

Catalog number: 425770250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 425770250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
68282-53-1
IUPAC Name
5-methyl-1H-imidazole-4-carbaldehyde
Molecular Formula
C5H6N2O
InChI Key
KMWCSNCNHSEXIF-UHFFFAOYSA-N
SMILES
CC1=C(C=O)N=CN1
NMR (1H-NMR)
Consistent with structure
Appearance (Color)
White to beige
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
168°C to 170°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text