Thermo Scientific Chemicals

6-Carboxyfluorescein, 96%

Catalog number: 425801000
100 mg, Glass bottle, Each
Thermo Scientific Chemicals

6-Carboxyfluorescein, 96%

Catalog number: 425801000
100 mg, Glass bottle, Each
Quantity
Packaging
Catalog number: 425801000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
3301-79-9
IUPAC Name
3',6'-dihydroxy-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthene]-6-carboxylic acid
Molecular Formula
C21H12O7
InChI Key
BZTDTCNHAFUJOG-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=C2C(=O)OC3(C2=C1)C1=CC=C(O)C=C1OC1=CC(O)=CC=C31
Infrared spectrum
Conforms
Appearance (Form)
Powder
NMR (1H-NMR)
Consistent with structure
HPLC
>=95 %
Appearance (Color)
Yellow to orange

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text