5-Carboxyfluorescein, 95%, Thermo Scientific Chemicals
5-Carboxyfluorescein, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Carboxyfluorescein, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
100 mg
Packaging:
Glass bottle
Catalog number 425815000
Price (USD)/ Each
-
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS76823-03-5
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
HPLC>=94 %
Appearance (Form)Powder
NMR (1H-NMR)Consistent with structure
Appearance (Color)Yellow to orange
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only