N-BOC-Pyrrolidine, 97%, Thermo Scientific Chemicals
N-BOC-Pyrrolidine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-BOC-Pyrrolidine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 425820250
Price (USD)/ Each
228.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
269.00 
Save 40.35 (15%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
228.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
269.00 
Save 40.35 (15%)
Add to cart
N-BOC-Pyrrolidine, 97%, Thermo Scientific Chemicals
Catalog number425820250
Price (USD)/ Each
228.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
269.00 
Save 40.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nametert-butyl pyrrolidine-1-carboxylate
Molecular FormulaC9H17NO2
InChI KeyLPQZERIRKRYGGM-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)N1CCCC1
Molecular Weight (g/mol)171.24
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
NMR (1H-NMR)Consistent with structure
Appearance (Form)Liquid
GC>=96.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only