Thermo Scientific Chemicals

Sulfur trioxide N,N-dimethylformamide complex, 47+% active SO3, Thermo Scientific Chemicals

Catalog number: 425850050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Sulfur trioxide N,N-dimethylformamide complex, 47+% active SO3, Thermo Scientific Chemicals

Catalog number: 425850050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 425850050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
29584-42-7
IUPAC Name
N,N-dimethylformamide; sulfonylideneoxidane
Molecular Formula
C3H7NO4S
InChI Key
AFDQGRURHDVABZ-UHFFFAOYSA-N
SMILES
O=S(=O)=O.CN(C)C=O
Appearance (Color)
White to beige to light brown
Infrared spectrum
Conforms
Titration with NaOH
>=47.0 % (as SO3)
Appearance (Form)
Powder or crystals

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text