Thermo Scientific Chemicals

1-Chlorobutane, 99.8%, for HPLC, Thermo Scientific Chemicals

Catalog number: 425880010
1 L, Glass bottle, Each
Thermo Scientific Chemicals

1-Chlorobutane, 99.8%, for HPLC, Thermo Scientific Chemicals

Catalog number: 425880010
1 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 425880010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
109-69-3
IUPAC Name
1-chlorobutane
Molecular Formula
C4H9Cl
InChI Key
VFWCMGCRMGJXDK-UHFFFAOYSA-N
SMILES
CCCCCl
GC
>=99.7 %
Residue after evaporation
=<2 ppm
UV
1 cm cell vs HPLC-grade water
Appearance (Form)
Liquid
Water
=<0.020 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLC
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text