1-Chlorobutane, 99.8%, for HPLC, Thermo Scientific Chemicals
1-Chlorobutane, 99.8%, for HPLC, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Chlorobutane, 99.8%, for HPLC, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass bottle
Catalog number 425880025
Price (USD)/ Each
255.65
Online exclusive
284.00 
Save 28.35 (10%)
-
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
255.65
Online exclusive
284.00 
Save 28.35 (10%)
Add to cart
1-Chlorobutane, 99.8%, for HPLC, Thermo Scientific Chemicals
Catalog number425880025
Price (USD)/ Each
255.65
Online exclusive
284.00 
Save 28.35 (10%)
-
Add to cart
Chemical Identifiers
CAS109-69-3
IUPAC Name1-chlorobutane
Molecular FormulaC4H9Cl
InChI KeyVFWCMGCRMGJXDK-UHFFFAOYSA-N
SMILESCCCCCl
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.7 %
Residue after evaporation=<2 ppm
UV1 cm cell vs HPLC-grade water
Appearance (Form)Liquid
Water=<0.020 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLC
RUO – Research Use Only